missing translation for 'onlineSavingsMsg'
Learn More

Metformin hydrochloride, MedChemExpress

Product Code. p-200096770
Encompass_Preferred
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 16477358

Brand: Medchem Express HY17471A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Metformin hydrochloride (1,1-Dimethylbiguanide hydrochloride) inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin hydrochloride triggers autophagy.

Metformin hydrochloride (1,1-Dimethylbiguanide hydrochloride) inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin hydrochloride triggers autophagy.

Specifications

Chemical Name or Material Metformin hydrochloride
Physical Form Solid
CAS 1115-70-4
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C4H12ClN5
For Use With (Application) Cancer-Kinase/protease
Synonym 1,1-Dimethylbiguanide hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (603.79 mM) ∣DMSO : ≥ 1.7 mg/mL (10.26 mM)
SMILES NC(NC(N(C)C)=N)=N.Cl
Molecular Weight (g/mol) 165.62
Formula Weight 165.62
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.