missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Metformin (Hydrochloride)

Product Code. p-7217516
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17150192

Brand: STEMCELL Technologies 73252

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AMPK activator; Mitochondrial respiratory chain complex I inhibitor

  • Metformin is an activator of the AMP-activated protein kinase (AMPK) pathway, and an inhibitor of mitochondrial respiratory chain complex 1 (Rena et al.; Viollet et al.) It acts as an antihyperglycemic agent to lower plasma glucose levels and improve insulin sensitivity (Viollet et al.) This product is provided as a hydrochloride salt of the molecule

Specifications

CAS 1115-70-4
Synonym Apophage; Diaformin; Fornidd; Glucoformin; Glucophage; LA 6023; Melbin; Orabet; Riomet; Walaphage
Quantity 1 g
Molecular Formula C4H11N5 · HCl
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.