missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Metformin (Hydrochloride)

Product Code. 17150192
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17150192 1 g 1g
17160192 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17150192 Supplier STEMCELL Technologies Supplier No. 73252
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

AMPK activator; Mitochondrial respiratory chain complex I inhibitor

  • Metformin is an activator of the AMP-activated protein kinase (AMPK) pathway, and an inhibitor of mitochondrial respiratory chain complex 1 (Rena et al.; Viollet et al.) It acts as an antihyperglycemic agent to lower plasma glucose levels and improve insulin sensitivity (Viollet et al.) This product is provided as a hydrochloride salt of the molecule

Specifications

CAS 1115-70-4
Synonym Apophage; Diaformin; Fornidd; Glucoformin; Glucophage; LA 6023; Melbin; Orabet; Riomet; Walaphage
Quantity 1 g
Molecular Formula C4H11N5 · HCl
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.