Learn More
Hanna Instruments™ Copper High Range Meter Reagents
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
Description
- High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements
- Follow the bicinchoninate method in which the reaction between copper and the reagent causes a color change in the sample
- Pre-made reagents for ease of use
- Prepared with high purity chemicals
- Reagents for the colorimetric determination of copper
Specifications
Specifications
| Quantity | 100 Pack |