missing translation for 'onlineSavingsMsg'
Learn More

Meteorin-like/METRNL Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30731324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731324 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731324 Supplier Novus Biologicals Supplier No. AF6679MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Meteorin-like/METRNL Polyclonal antibody specifically detects Meteorin-like/METRNL in Mouse samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Meteorin-like/METRNL
Applications Immunohistochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias meteorin, glial cell differentiation regulator-like, meteorin-like protein, MGC99788
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Meteorin-like/METRNL, Accession # Q8VE43
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284207
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.