missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meteorin-like/METRNL Antibody (829535) [DyLight 350], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB66791D
This item is not returnable.
View return policy
Description
Meteorin-like/METRNL Monoclonal antibody specifically detects Meteorin-like/METRNL in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Meteorin-like/METRNL | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284207 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 829535 | |
| Flow Cytometry, CyTOF-ready | |
| meteorin, glial cell differentiation regulator-like, meteorin-like protein, MGC99788 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse METRNL, Gln46-Glu311, Accession # Q8VE43 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction