missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meteorin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | Meteorin |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18462011
|
Novus Biologicals
NBP2-14231-25ul |
25ul |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18045549
|
Novus Biologicals
NBP2-14231 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Meteorin Polyclonal specifically detects Meteorin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Meteorin | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| C16orf23, c380A1.2 | |
| METRN | |
| IgG | |
| Affinity Purified | |
| Specificity of human Meteorin antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 79006 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: VTHDVELQESVITVVAARVLRQTPPLFQAGRSGDQGLTSIRTPLRCGVHPGPGTFLFMGWSRFG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts