missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metenolone enanthate, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Metenolone enanthate
Specifications
Specifications
| Chemical Name or Material | Primobolan |
| CAS | 303-42-4 |
| Analyte or Component Names | Primobolan |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H42 O3 |
| Synonym | Metenolone enanthate, Methenolone 17-enanthate, Methenolone enanthate, NSC 64967, Primobolan Depot, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCCCCCC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CC[C@H]4 CC(=O)C=C(C)[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | [(5 S,8 R,9 S,10 S,13 S,14 S,17 S)-1,10,13-trimethyl-3-oxo-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoate |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction