missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metazachlor-sulfinyl-acetic acid BH 479-9 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Metazachlor-sulfinyl-acetic acid BH 479-9 |
| CAS | 1246215-97-3 |
| Analyte or Component Names | Metazachlor-sulfinyl-acetic acid BH 479-9 |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 N3 O4 S |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Metazachlor-sulfinyl-acetate, 479M09, BH 479-9, Acetic acid, 2-[[2-[(2,6-dimethylphenyl)(1H-pyrazol-1-ylmethyl)amino]-2-oxoethyl]sulfinyl]- |
| InChI Formula | InChI=1S/C16H19N3O4S/c1-12-5-3-6-13(2)16(12)19(11-18-8-4-7-17-18)14(20)9-24(23)10-15(21)22/h3-8H,9-11H2,1-2H3,(H,21,22) |
| SMILES | Cc1cccc(C)c1N(Cn2cccn2)C(=O)CS(=O)CC(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?