missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Monoclonal Antibody (HMPV123), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516713
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Mouse anti Metapneumovirus antibody, clone HMPV123 recognizes human metapneumovirus (hMPV), a member of the Paramyxoviridae RNA virus family.
Metapneumovirus Monoclonal antibody specifically detects Metapneumovirus in Virus samples. It is validated for Immunocytochemistry.Specifications
Metapneumovirus | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry | |
HMPV123 | |
Unconjugated | |
h h h-MPV; MPV | |
Sub-type A hMPV virus isolate NCL03-4/145 | |
200 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |