Learn More
Metapneumovirus Antibody (hMPV57) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-53105
 View more versions of this product
Product Code. 18240581
  • £348.00 / 0.10mL
Description
Metapneumovirus Monoclonal specifically detects Metapneumovirus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Metapneumovirus
Monoclonal
1.0 mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Mouse
Protein A purified
RUO
Primary
Virus
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
hMPV57
Unconjugated
PBS with 0.02% Sodium Azide
Human MPV strain NCL-174
0.1 mL
Virology Bacteria and Parasites
Identifies Metapneumovirus nucleoprotein of both types A and B.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1