missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ METAP1D Polyclonal Antibody

Product Code. 15764341 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15764341 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15764341 Supplier Invitrogen™ Supplier No. PA556433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VGNVDECGKK LVEVARRCRD EAIAACRAGA PFSVIGNTIS HITHQNGFQV CPHFVGHGIG SYFHGHPEIW HHA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.

The N-terminal methionine excision pathway is an essential process in which the N-terminal methionine is removed from many proteins, thus facilitating subsequent protein modification. In mitochondria, enzymes that catalyze this reaction are celled methionine aminopeptidases (MetAps, or MAPs; EC 3.4.11.18).
TRUSTED_SUSTAINABILITY

Specifications

Antigen METAP1D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene METAP1D
Gene Accession No. Q6UB28
Gene Alias 2310066F24Rik; 3110033D18Rik; AV117938; CDS of metAP-3 within PCR fragment; MAP 1D; Map1d; MetAP 1D; METAP1D; Metap1l; Metapl1; methionine aminopeptidase 1D, mitochondrial; methionine aminopeptidase-like 1; methionyl aminopeptidase type 1D (mitochondrial); methionyl aminopeptidase type 1D, mitochondrial; Mnpepl; Peptidase M 1D
Gene Symbols METAP1D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human METAP1D. Recombinant protein control fragment (Product #RP-103821).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254042
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.