missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metanil Yellow, TRC
Specifications
Specifications
| Chemical Name or Material | Acid Yellow 36 |
| CAS | 587-98-4 |
| Molecular Formula | C18 H14 N3 O3 S . Na |
| Quantity | 500 mg |
| Synonym | Benzenesulfonic acid','3-[[4-(phenylamino)phenyl]azo]-','monosodium salt (9CI), Benzenesulfonic acid','m-(p-anilinophenylazo)-','sodium salt (6CI), C.I. Acid Yellow 36 (7CI), C.I. Acid Yellow 36','monosodium salt (8CI), 11363 Yellow, Acid Gold Yellow G, Acid Golden G, Acid Golden Yellow G, Acid Leather Yellow PRW, Acid Leather Yellow R, Acid Metanil Yellow G, Acid Metanil Yellow MT, Acid Yellow 36, Acid Yellow Metanil, Acid Yellow Metanilic, Aizen Metanil Yellow, Amacid Yellow M, BTK Metanil Yellow, Basacid Yellow 230, Basacid Yellow 232, Basacid Yellow 234, Borunil Yellow A-G, Brasilan Metanil Yellow, Bucacid Metanil Yellow, C Ext. Yellow 10, C.I. 13065, Calcocid Yellow MXXX, Cogilor Yellow 114.12, Colorosacid Yellow Methanilic, Conacid Yellow CK, Diacid Metanil Yellow, Dinacid Metanil Yellow, Duasyn Acid Yellow C 01, Dyacid Yellow M, Dycosacid Golden Yellow G, Dycosacid Metanil Yellow MT, Dycosacid Yellow MT, Egacid Yellow M, Egacid Yellow MA, Eniacid Metanil Yellow GN, Ext D & C Yellow 1, Ext D and C Yellow No. 1, Fabracid Yellow S-M, Fenazo Yellow M, Hidacid Metanil Yellow, Hispacid Yellow MG, Ichoacid Metanil Yellow R, Indacid Metanil Yellow, Japan Yellow 406, Japan Yellow No. 406, Java Metanil Yellow G, KI 406, Kemacid Metanil Yellow R, Kiton Orange MNO, Kiton Yellow MS, Lowalan Yellow 36, Metanil Yellow, Metanil Yellow 1955, Metanil Yellow 205, Metanil Yellow 216, Metanil Yellow 222, Metanil Yellow 23035, Metanil Yellow C, Metanil Yellow E, Metanil Yellow Extra, Metanil Yellow F, Metanil Yellow G, Metanil Yellow Griesbach, Metanil Yellow K, Metanil Yellow KRSU, Metanil Yellow M, Metanil Yellow M 3X, Metanil Yellow O, Metanil Yellow PL, Metanil Yellow R, Metanil Yellow S, Metanil Yellow Supra P, Metanil Yellow VS, Metanil Yellow WS, Metanil Yellow Y, Metanil Yellow YK, Metanile Yellow O, Metanilic Yellow, Methanil Yellow G, Methanyl yellow, Mitsui Metanil Yellow, Multacid Yellow M, Multicuer Yellow AM, Pacid Mentanil Yellow BG, Paper Metanil Yellow P, Remaderm Yellow HPR, Ritacid Metanil Yellow, Rybacel Yellow AM, Sandolan Yellow E-MY, Shikiso Metanil Yellow, Sodium 4'-anilinoazobenzene-3-sulfonate, Sodium m-(4-anilinophenylazo)benzenesulfonate, Sodium m-sulfonate-p-phenylazodiphenylamine, Solcoderm Yellow FM, Symulon Metanil Yellow, Takaoka Metanil Yellow, Tertracid Yellow M, Triacid Metanil Yellow, Triacid Metanil Yellow M, Tropaeolin G, Vicoacid Metanil Yellow, Vondacid Metanil Yellow G, Vopsider Metanil Yellow, Water Golden Yellow 028431, Yellow 406, Yellow Methanil, Yellow No. 406, Yodochrome Metanil Yellow |
| InChI Formula | InChI=1S/C18H15N3O3S.Na/c22-25(23,24)18-8-4-7-17(13-18)21-20-16-11-9-15(10-12-16)19-14-5-2-1-3-6-14;/h1-13,19H,(H,22,23,24);/q;+1/p-1 |
| SMILES | [Na+].[O-]S(=O)(=O)c1cccc(c1)N=Nc2ccc(Nc3ccccc3)cc2 |
| IUPAC Name | sodium;3-[(E)-(4-anilinophenyl)diazenyl]benzenesulfonate |
| Molecular Weight (g/mol) | 375.38 |
| Formula Weight | 375.0654 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?