missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Metanil Yellow (Tech.), 85%

Product Code. 10771252 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
Product Code. Quantity unitSize
10771252 250 g 250g
10472971 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 10771252

Brand: Thermo Scientific Chemicals 413722500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 587-98-4 | C18H14N3NaO3S | 375.378 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-98-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.378
InChI Key NYGZLYXAPMMJTE-UHFFFAOYSA-M
Synonym Acid Yellow 36, 3-[[4-(Phenylamino)phenyl]azo]benzenesulfonic acid monosodium salt, C.I. 13065, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
PubChem CID 3935589
ChEBI CHEBI:87235
IUPAC Name sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]

Specifications

Dye Content 80% min.
Beilstein 16, 330
Merck Index 15, 5999
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol, moderately soluble in benzene and ether, slightly soluble in acetone
Infrared Spectrum Authentic
Formula Weight 375.38
Grade Pure
Packaging Glass bottle
Color Orange
Quantity 250 g
Physical Form Powder
Chemical Name or Material Metanil Yellow, tech., 85%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.