missing translation for 'onlineSavingsMsg'
Learn More

Metanil Yellow, MP Biomedicals

Product Code. 11427575 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427575 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11427575 Supplier MP Biomedicals Supplier No. 0521194280
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 587-98-4
Molecular Formula C18H14N3NaO3S
Quantity 100 g
Synonym metanil yellow, acid yellow 36, acid metanil yellow, monoazo, amacid yellow m, fenazo yellow m, acid golden g, kiton yellow ms, metanil yellow c, metanil yellow e
InChI Key NYGZLYXAPMMJTE-UHFFFAOYSA-M
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
Molecular Weight (g/mol) 375.378
PubChem CID 3935589
ChEBI CHEBI:87235
Formula Weight 375.38
Chemical Name or Material Metanil Yellow
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.