missing translation for 'onlineSavingsMsg'
Learn More

Metamizole Sodium Monohydrate, Mikromol™

Product Code. 17961626
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17961626

Brand: Mikromol MM0052.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Metamizole Sodium Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Metamizole Sodium Monohydrate
CAS 5907-38-0
Analyte or Component Names Metamizole Sodium Monohydrate
Shipping Condition Room Temperature
Molecular Formula C13 H16 N3 O4 S . Na . H2 O
Certifications/Compliance ISO 17034
Synonym Metamizole Sodium Monohydrate, 1-[(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-methanesulfonic acid monosodium salt monohydrate, (Antipyrinylmethylamino)-methanesulfonic acid sodium salt monohydrate, [(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-methanesulfonic acid sodium salt monohydrate, Dipyrone monohydrate, Metamizol monohydrate, Sodium (antipyrinylmethylamino)methanesulfonate hydrate
InChI Formula InChI=1S/C13H17N3O4S.Na.H2O/c1-10-12(14(2)9-21(18,19)20)13(17)16(15(10)3)11-7-5-4-6-8-11;;/h4-8H,9H2,1-3H3,(H,18,19,20);;1H2/q;+1;/p-1
SMILES O.[Na+].CN(CS(=O)(=O)[O-])C1=C(C)N(C)N(C1=O)c2ccccc2
IUPAC Name sodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-methylamino]methanesulfonate;hydrate
Molecular Weight (g/mol) 351.35
Quantity 250 mg
Formula Weight 351.0865 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.