missing translation for 'onlineSavingsMsg'
Learn More

Metamizole Sodium Monohydrate 1.0 mg/ml in Dimethyl Sulfoxide (as anhydrous free acid), LoGiCal

Product Code. 30459223
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30459223

Brand: LoGiCal LGCAMP0052.0029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Metamizole Sodium Monohydrate 1.0 mg/ml in Dimethyl Sulfoxide (as anhydrous free acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Metamizole Sodium Monohydrate
CAS 5907-38-0
Analyte or Component Names Metamizole Sodium Monohydrate
Shipping Condition Ice Pack (-20°C)
Molecular Formula C13 H16 N3 O4 S . Na . H2 O
Certifications/Compliance ISO 17034
Synonym Metamizole Sodium Monohydrate, 1-[(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1 H-pyrazol-4-yl)methylamino]-methanesulfonic acid monosodium salt monohydrate, (Antipyrinylmethylamino)-methanesulfonic acid sodium salt monohydrate, [(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1 H-pyrazol-4-yl)methylamino]-methanesulfonic acid sodium salt monohydrate, Dipyrone monohydrate, Metamizol monohydrate, Sodium (antipyrinylmethylamino)methanesulfonate hydrate
InChI Formula InChI=1 S/C13H17N3O4S.Na.H2O/c1-10-12(14(2)9-21(18,19)20)13(17)16(15(10)3)11-7-5-4-6-8-11;;/h4-8 H,9H2,1-3H3,(H,18,19,20);;1H2/q;+1;/p-1
SMILES O.[Na+].CN(CS(=O)(=O)[O-])C1=C(C)N(C)N(C1=O)c2ccccc2
IUPAC Name sodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-methylamino]methanesulfonate;hydrate
Solvent or Matrix Dimethyl Sulfoxide
Industry Type Forensics
Molecular Weight (g/mol) 351.35
Quantity 1 mL
Formula Weight 351.0865 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.