missing translation for 'onlineSavingsMsg'
Learn More

Metamizole sodium hydrate, MedChemExpress

Product Code. 16432266
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432266 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16432266 Supplier Medchem Express Supplier No. HYB1279
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Metamizole sodium hydrate is a potent analgesic drug that has been demonstrated to inhibit cyclooxygenase (COX)[1].

Metamizole sodium hydrate is a potent analgesic drug that has been demonstrated to inhibit cyclooxygenase (COX)[1].

Specifications

Chemical Name or Material Metamizole sodium hydrate
CAS 5907-38-0
Color White
Purity Grade Notes Research
Molecular Formula C13H18N3NaO5S
Quantity 500 mg
Solubility Information DMSO : 100 mg/mL (284.62 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (284.62 mM)
SMILES O=S(CN(C1=C(C)N(C)N(C2=CC=CC=C2)C1=O)C)(O[Na])=O.O
Molecular Weight (g/mol) 351.35
Formula Weight 351.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.