missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Metallothionein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577772
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of PA5-77772 was sufficient for detection of Metallothionein in 15 μg of pseudomonas aeruginosa purified Metallothionein (pmtA-GST) protein by ECL immunoblot analysis using goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 36 kDa.
Metallothioneins (MTs) are a family of low molecular weight, heavy metal-binding proteins characterized by a high cysteine content and lack of aromatic amino acids. MTs bind 7 to 12 heavy metal atoms per molecule of protein. They are ubiquitous in the animal and plant kingdoms and are also found in prokaryotes. In mammals, the cysteine residues are absolutely conserved and serve to coordinate heavy metal atoms such as zinc, cadmium, and copper via mercaptide linkages.
Specifications
| Metallothionein | |
| Polyclonal | |
| Unconjugated | |
| Mt1 | |
| Mt1, PA2140 | |
| Synthetic peptide from the mid-protein of Pseudomonas aeruginosa Metallothionein. | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| CES 1; CES1; LOC100343802; LOC100630794; Metallothionein; metallothionein 1; metallothionein 1a; metallothionein 1A (functional); metallothionein 1S; Metallothionein 2; Metallothionein 2A; Metallothionein IA; Metallothionein II; metallothionein-1; Metallothionein-1A; metallothionein-1A-like; Metallothionein2; metallothionein-I; Metallothionein-IA; MGC32848; Mt; MT 1A; MT 2; MT 2A; MT IA; MT II; Mt1; MT-1; MT1A; MT-1A; MT1S; MT2; MT2A; MTC; MT-I; MT-IA; Probable pmtA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20220066, 880550 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur