missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Metallothionein Monoclonal Antibody (UC1MT)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA125479
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
A band of approximately 15 kDa is detected. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Metallothioneins (MTs) are a family of low molecular weight, heavy metal-binding proteins characterized by a high cysteine content and lack of aromatic amino acids. MTs bind 7 to 12 heavy metal atoms per molecule of protein. They are ubiquitous in the animal and plant kingdoms and are also found in prokaryotes. In mammals, the cysteine residues are absolutely conserved and serve to coordinate heavy metal atoms such as zinc, cadmium, and copper via mercaptide linkages.
Tekniske data
| Metallothionein | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.2 | |
| P02802, P02803, P04731 | |
| LOC100630794, Mt1, MT1A | |
| Full length protein (cross linked rabbit liver MT1 and MT2). | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Horse, Fish | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| UC1MT | |
| Unconjugated | |
| MT1A | |
| CES 1; CES1; LOC100343802; LOC100630794; Metallothionein; metallothionein 1; metallothionein 1a; metallothionein 1A (functional); metallothionein 1S; Metallothionein 2; Metallothionein 2A; Metallothionein IA; Metallothionein II; metallothionein-1; Metallothionein-1A; metallothionein-1A-like; Metallothionein2; metallothionein-I; Metallothionein-IA; MGC32848; Mt; MT 1A; MT 2; MT 2A; MT IA; MT II; Mt1; MT-1; MT1A; MT-1A; MT1S; MT2; MT2A; MTC; MT-I; MT-IA; Probable pmtA | |
| Mouse | |
| Protein G | |
| RUO | |
| 100630794, 17748, 24567, 4489 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion