Learn More
Invitrogen™ Metallothionein Monoclonal Antibody (1F5), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545896
Description
A 1:1,000 dilution of MA5-45896 was sufficient for detection of Metallothionein in 1 μg of purified Pseudomonas aeruginosa Metallothionein (PmtA)-GST tagged protein by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects 36 kDa band corresponding to Pseudomonas aeruginosa Metallothionein.
Metallothioneins (MTs) are a family of low molecular weight, heavy metal-binding proteins characterized by a high cysteine content and lack of aromatic amino acids. MTs bind 7 to 12 heavy metal atoms per molecule of protein. They are ubiquitous in the animal and plant kingdoms and are also found in prokaryotes. In mammals, the cysteine residues are absolutely conserved and serve to coordinate heavy metal atoms such as zinc, cadmium, and copper via mercaptide linkages.
Specifications
| Metallothionein | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| 0 | |
| Mt1, PA2140 | |
| Recombinant full length Pseudomonas aeruginosa Metallothionein protein. | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 1F5 | |
| FITC | |
| Mt1 | |
| CES 1; CES1; LOC100343802; LOC100630794; Metallothionein; metallothionein 1; metallothionein 1a; metallothionein 1A (functional); metallothionein 1S; Metallothionein 2; Metallothionein 2A; Metallothionein IA; Metallothionein II; metallothionein-1; Metallothionein-1A; metallothionein-1A-like; Metallothionein2; metallothionein-I; Metallothionein-IA; MGC32848; Mt; MT 1A; MT 2; MT 2A; MT IA; MT II; Mt1; MT-1; MT1A; MT-1A; MT1S; MT2; MT2A; MTC; MT-I; MT-IA; Probable pmtA | |
| Mouse | |
| Protein G | |
| RUO | |
| 20220066, 880550 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.