missing translation for 'onlineSavingsMsg'
Learn More
Learn More
metallothionein Antibody (N11-G), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-79083-50UL
This item is not returnable.
View return policy
Description
metallothionein Monoclonal specifically detects metallothionein in Human, Rabbit samples. It is validated for Western Blot, ELISA.
Specifications
| metallothionein | |
| Monoclonal | |
| Unconjugated | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| NBP1-79083 immunogen is identical and covers isoforms#: 1A P04731, #1B P07438, #1E P04732,#1F P04733, #1G P13640, #1H P80294, #1I P80295, #1L Q93083, #1M Q8N339, #1Q Q86YX0, #1S Q86YX5, #1Y Q8TDC4, #2A P18055. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| N11-G | |
| Western Blot 1:1000, ELISA 1:5000-1:10000 | |
| Peptide corresponding to the N-terminal sequence of human metallothionein (covering isoforms 1A, B, E, F, G, H, I, L, M, Q, S, Y, 2A) | |
| 50 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction