missing translation for 'onlineSavingsMsg'
Learn More

metallothionein Antibody (1F5), Novus Biologicals™

Product Code. 18346423
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346423 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346423 Supplier Bio-Techne Supplier No. NBP318261

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

metallothionein Monoclonal antibody specifically detects metallothionein in Bacteria samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen metallothionein
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1F5
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:800
Formulation PBS pH 7.4, 50% glycerol
Host Species Mouse
Immunogen Recombinant full length Pseudomonas aeruginosa Metallothionein protein
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.