missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Metallothionein-3 Polyclonal specifically detects Metallothionein-3 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Metallothionein-3 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence validated for ICC/IF from a verified customer review., Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | P25713 |
| Gene Alias | GIFBmetallothionein-3, GIFGRIF, Growth inhibitory factor, metallothionein 3, metallothionein 3 (growth inhibitory factor (neurotrophic)), metallothionein-III, MT-3, MT-III |
| Gene Symbols | MT3 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?