missing translation for 'onlineSavingsMsg'
Learn More

Metalloids/Hydride Elements Standard: As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 μg/mL in 20% HCl, tr. HF, VHG™

Product Code. 30003030
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003030 500 mL 500mL
17997289 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30003030 Supplier VHG Labs Supplier No. VHGSM50B500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Metalloids/Hydride Elements Standard: As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 μg/mL in 20% HCl, tr. HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Germanium/Antimony/Thallium/Tin/Selenium/Gallium/Indium/Tellurium/Bismuth/Lead/Arsenic
CAS 7440-56-4, 7440-36-0, 7440-28-0, 7440-31-5, 7782-49-2, 7440-55-3
Methods ICP/ICP-MS
Analyte or Component Names Germanium/Antimony/Thallium/Tin/Selenium/Gallium/Indium/Tellurium/Bismuth/Lead/Arsenic
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HCl
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Germanium 99.97 μg/mL/Antimony 99.98 μg/mL/Thallium 99.98 μg/mL/Tin 99.98 μg/mL/Selenium 99.99 μg/mL/Gallium 100 μg/mL/Indium 100 μg/mL/Tellurium 100 μg/mL/Bismuth 100.1 μg/mL/Lead 100.1 μg/mL/Arsenic 100.2 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.