missing translation for 'onlineSavingsMsg'
Learn More

Metaldehyde, 98%

Product Code. 11478156
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478156 250 g 250g
11468156 50 g 50g
11458156 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478156 Supplier Thermo Scientific Alfa Aesar Supplier No. A15292.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-62-3 | C8H16O4 | 176.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 108-62-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.212
MDL Number MFCD00071549
InChI Key GKKDCARASOJPNG-UHFFFAOYSA-N
Synonym metaldehyde, metacetaldehyde, cekumeta, ariotox, halizan, metason, acetaldehyde, tetramer, slug-tox, corry's slug death, metaldehyd
PubChem CID 61021
ChEBI CHEBI:81931
IUPAC Name 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
SMILES CC1OC(OC(OC(O1)C)C)C

Especificaciones

Melting Point 192°C to 196°C (sublimation)
Flash Point 55°C (131°F)
Quantity 250 g
UN Number UN1332
Sensitivity Light sensitive
Merck Index 14,5923
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material Metaldehyde

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.