missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Metal Springs for Vial Inserts

Product Code. 10185161 Shop All Macherey-Nagel Products
Change view
Click to view available options
Height (Metric):
30 mm
40 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
10185161 40 mm Pack of 100
10226311 30 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10185161 Supplier Macherey-Nagel™ Supplier No. 702974
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use with micro inserts for autosampler vials. Macherey-Nagel™ Metal Springs for Vial Inserts are optional metal springs that act as a shock absorber to prevent breakage when inserting or using a compatible inserts.

  • Spring 702974: Use with 0.3mL insert for N13 screw neck vials (mfr. no. 702972)
  • Spring 702974.1: Use with 0.1mL insert (mfr. no. 702968) or 0.15mL insert (mfr. no. 702968.1) for small opening vials
  • Metal spring protects against damage during insertion or when a needle presses down on the bottom
  • Strong, flexible springs act as a shock absorber
  • Packaged as 100 pieces in a polyethylene (PE) bag
  • Compatible inserts and vials sold separately

Specifications

Material Metal
Quantity 100 Pk.
For Use With (Equipment) 0.3 mL Insert for N13 Screw Top Vial (702972)
Height (Metric) 40 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.