missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS Metal Lysing Tube with Grinding Ball

Product Code. 30690121 Shop All MP Biomedicals Products
Change view
Click to view available options
Capacity:
50 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
30690121 50 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30690121 Supplier MP BIOMEDICALS Supplier No. 116957001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for grinding, lysing, and homogenizing your most resistant samples. These tubes are machined from titanium stock, which provides much greater strength than conventional stainless steel and aluminum tubes and allows for unlimited use.

Titanium's strength is comparable to steel however its weight is 40% lower. Constructed from titanium, these tubes and grinding matrix are tough enough to stand up to the most demanding mechanical punishment that can cause traditional thermoplastic tubes to crack. Our tubes are machined from premium grade billet and deliver superior strength over less expensive production methods. An oblique angle conical bottom provides a better impact surface than the rounded bottoms of the deep-drawn tube. The titanium threaded cap provides a leak-proof closure without the energy-robbing alternatives like plastic flange screw caps or rubber stoppers. A Teflon O-ring prevents leakage and can be cleaned with detergent and/or autoclaving or replaced entirely between samples. Machined straight knurls on the cap provide a firm grip for easy opening and closing. Research areas and sample type include:

  • Environmental and Agriculture: Tough seeds such as dried corn, soybeans, wheat, tomato; wood, bark, roots, animal claws and hooves
  • Forensics: Bone, teeth, hair, fingernails, and non-biological substrates
  • Cancer and Other Diseases: Tough tissues, bone, cartilage, and skin
  • Industrial: Non-biological, rocks and minerals, plastics and composites, printed circuit boards, wood and building materials

Application Notes

  • Dry grinding of very tough or hard samples in which generated heat can damage plastic tubes
  • Cryogenic dry grinding where severe cold temperatures (dry ice or liquid nitrogen) can damage plastic tubes
  • Milling or grinding non-biological samples where plastic contamination is of concern
  • Sample processing with solvents or chemicals that are incompatible with plastics.

Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 50 mL
Isolation Technology Beads
Product Type Lysing tube
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.