missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Metal Lysing Tube, 2 mL, with 0.75 x 0.5 in. Grinding Cylinder

Product Code. 15808674 Shop All MP Biomedicals Products
Click to view available options
Quantity:
2 Pk.
3 Pk.
6 Pk.
Unit Size:
Pack of 2
Pack of 3
Pack of 6
This item is not returnable. View return policy

Product Code. 15808674

Brand: MP Biomedicals™ 116992002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Stainless steel lysing matrix tube with the grinding cylinder is ideal for grinding, lysis, and homogenization of resistant samples.

This stainless steel lysing matrix tube with the grinding cylinder is ideal for grinding, lysis, and homogenization of resistant samples. Constructed from 308 SS, these tubes and grinding cylinders are tough enough to stand up to the most demanding mechanical punishment that can cause traditional thermoplastic tubes to crack. The tubes are machined from premium grade billet and deliver superior strength over less expensive production methods such as deep-drawn aluminum tubes. An oblique angle conical bottom provides a better impact surface than the rounded bottoms of deep-drawn tubes. The stainless steel threaded cap provides a leak-proof closure without the energy-robbing alternatives like plastic flange screw caps or rubber stoppers. A Teflon O-ring prevents leakage, and can be cleaned with detergent and/or autoclaving, or replaced entirely between samples. Machined knurls on the cap provide a firm grip to easy opening and closing.

Sample types include:

  • Tough seeds such as dried corn, soybeans, wheat, tomato, and Chile
  • Wood, bark, roots, animal claws and hooves
  • Bone, teeth, hair, fingernails, and non-biological substrates
  • Tough tissues, bone, cartilage, skin
  • Non-biological, rocks and minerals, plastics and composites, printed circuit boards, wood, and building materials
Application Notes
  • Dry grinding of very tough or hard samples in which generated heat can damage plastic tubes.
  • Cryogenic dry grinding where severe cold temperatures (dry ice or liquid nitrogen) can damage plastic tubes.
  • Milling or grinding non-biological samples where plastic contamination is of concern.
  • Sample processing with solvents or chemicals that are incompatible with plastics.
TRUSTED_SUSTAINABILITY

Specifications

Bead Type Stainless Steel
Sample Type Seeds, Wood, Bark, Roots, Bone, Teeth, Tissues, Cartilage, Rocks, Minerals, Plastics, Composites
Includes 0.75 x 0.5 in. grinding cylinder
Product Type Metal Lysing Tube
Quantity 2 Pk.

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.