Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 2, 3 or 4
Product Code. 11502355
Change view
Click to view available options
:
Gauge: 31
Product Code.
11502355 Gauge: 31
1 options
Product Code. 11502355 Supplier Hamilton™ Supplier No. 774817
Only null left
Add to basket
Add to basket
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Needle Point Number Point 2, 3 or 4
Quantity 6 Pk.