Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 2, 3 or 4
Product Code. 11512345
Change view
Click to view available options
Needle Gauge:
20 G
Unit Size:
Pack of 6
Product Code. Needle Gauge unitSize
11512345 20 G Pack of 6
1 options
Product Code. 11512345 Supplier Hamilton™ Supplier No. 774806
Only null left
Add to basket
Add to basket
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Autoclavable Autoclavable
Material Nickel-plated Brass (Hub), Stainless Steel
Needle Gauge 20 G
Diameter (Metric) Inner 0.60 mm
Diameter (Metric) Outer 0.91 mm
Product Type Luer Lock Custom Needle
Quantity 6/Pk.
Length (Metric) Needle 9.52 to 304 mm
Needle Point Style 2, 3, or 4
For Use With (Equipment) TLL Syringes and LT or TLL Connectors
Connection Type Standard Tapered