missing translation for 'onlineSavingsMsg'
Learn More

Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17992096
Change view
Click to view available options
Quantity:
50 g
100 g
200 g
400 g
Unit Size:
100g
200g
400g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992096 200 g 200g
17902276 50 g 50g
17940736 400 g 400g
17960606 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17992096 Supplier VHG Labs Supplier No. VHGMA51000200G
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium/Calcium/Magnesium/Phosphorus/Zinc
CAS 7440-39-3, 7440-70-2, 7439-95-4, 7723-14-0, 7440-66-6
Methods ICP/XRF/RDE
Analyte or Component Names Barium/Calcium/Magnesium/Phosphorus/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 75 cSt Hydrocarbon Oil
Quantity 200 g
Concentration or Composition (by Analyte or Components) Barium 1000 μg/g/Calcium 1000 μg/g/Magnesium 1000 μg/g/Phosphorus 1000 μg/g/Zinc 1000 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.