missing translation for 'onlineSavingsMsg'
Learn More

Metadoxine, MedChemExpress

Product Code. 16434637
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434637 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16434637 Supplier Medchem Express Supplier No. HYB1898

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Metadoxine blocks adipocyte differentiation in association with inhibition of the protein kinase A-cAMP response element binding protein (PKA-CREB) pathway.

Metadoxine blocks adipocyte differentiation in association with inhibition of the protein kinase A-cAMP response element binding protein (PKA-CREB) pathway.

Specifications

Chemical Name or Material Metadoxine
CAS 74536-44-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H18N2O6
Quantity 50 mg
Solubility Information H2O : ≥ 155 mg/mL (519.63 mM)
SMILES O=C(O)[C@H](CC1)NC1=O.OCC2=C(CO)C(O)=C(C)N=C2
Molecular Weight (g/mol) 298.29
Formula Weight 298.29
Percent Purity 99.38%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.