missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Metadherin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 406500
This item is not returnable.
View return policy
Description
Metadherin Polyclonal Antibody for ICC/IF, IHC (P), IP, Western Blot
Metadherin (Metastasis adhesion protein), also known as MTDH, LYsine-RIch CEACAM1 co-isolated (LYRIC), is a novel protein that localizes with the tight junction proteins ZO-1 and occludin in polarized epithelial cells. At the tight junction, it acts not as a structural component, but is rather recruited during the maturation of the tight junction complex. Metadherin is overexpressed in breast cancer tissue and breast tumor xenografts, while much lower levels are expressed in normal breast tissue. Metadherin binds to lung vasculature, one of the four common sites of breast cancer metastasis, through a C-terminal segment in the extracellular domain; blocking this lung-homing domain with antibodies or inhibiting metadherin with siRNA has been reported to inhibit breast cancer metastasis.
Specifications
| Metadherin | |
| Polyclonal | |
| Unconjugated | |
| MTDH | |
| 2610103J23Rik; 3D3; 3D3/Lyric; AEG1; AEG-1; astrocyte elevated gene 1; astrocyte elevated gene-1 protein; AV353288; D8Bwg1112e; LYRIC; LYRIC/ MTDH; LYRIC/3D3; lysine-rich CEACAM1 co-isolated protein; Metadherin; metastasis adhesion protein; MTDH; Protein LYRIC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 612147, 67154, 92140 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Q80WJ7, Q86UE4 | |
| MTDH | |
| Synthetic peptide derived from an internal region of the human, chimpanzee, mouse, and bovine metadherin (Mtdh, metastasis adhesion protein, LYsine-RIch CEACAM1 co-isolated (LYRIC), LYRIC/3D3, astrocyte elevated gene-1 (AEG-1)) protein, which differs from rat by one amino acid. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction