Läs mer
Invitrogen™ Metadherin Monoclonal Antibody (2F11C3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515564
Beskrivning
MA5-15564 targets Metadherin in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human samples. The MA5-15564 immunogen is purified recombinant fragment of human Metadherin expressed in E. Coli. MA5-15564 detects Metadherin which has a predicted molecular weight of approximately 64kDa.
Metadherin (Metastasis adhesion protein), also known as MTDH, LYsine-RIch CEACAM1 co-isolated (LYRIC), is a novel protein that localizes with the tight junction proteins ZO-1 and occludin in polarized epithelial cells. At the tight junction, it acts not as a structural component, but is rather recruited during the maturation of the tight junction complex. Metadherin is overexpressed in breast cancer tissue and breast tumor xenografts, while much lower levels are expressed in normal breast tissue. Metadherin binds to lung vasculature, one of the four common sites of breast cancer metastasis, through a C-terminal segment in the extracellular domain; blocking this lung-homing domain with antibodies or inhibiting metadherin with siRNA has been reported to inhibit breast cancer metastasis.
Specifikationer
| Metadherin | |
| Monoclonal | |
| Conc. Not Determined | |
| PBS with 0.03% sodium azide | |
| Q86UE4 | |
| MTDH | |
| Purified recombinant fragment of human Metadherin expressed in E. Coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2F11C3 | |
| Unconjugated | |
| MTDH | |
| 2610103J23Rik; 3D3; 3D3/Lyric; AEG1; AEG-1; astrocyte elevated gene 1; astrocyte elevated gene-1 protein; AV353288; D8Bwg1112e; LYRIC; LYRIC/ MTDH; LYRIC/3D3; lysine-rich CEACAM1 co-isolated protein; Metadherin; metastasis adhesion protein; MTDH; Protein LYRIC | |
| Mouse | |
| Protein G | |
| RUO | |
| 92140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.