missing translation for 'onlineSavingsMsg'
Learn More

Metadherin Antibody [PerCP], Novus Biologicals™

Product Code. 30735450 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735450 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735450 Supplier Novus Biologicals Supplier No. AF7180C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Metadherin Polyclonal antibody specifically detects Metadherin in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Metadherin
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias 3D3, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, protein LYRIC
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Metadherin, Lys167-Ser297, Accession # Q80WJ7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 92140
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.