missing translation for 'onlineSavingsMsg'
Learn More

Tetraconazole in Methanol 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12940164 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12940164

Brand: Fisher Chemical P853570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 112281-77-3
Physical Form Liquid
Molecular Formula C13H11Cl2F4N3O
Concentration 100 μg/mL
UN Number 1230
InChI Key LQDARGUHUSPFNL-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC(=C(C=C1Cl)Cl)C(CN2C=NC=N2)COC(C(F)F)(F)F
IUPAC Name 1-[2-(2,4-dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]-1,2,4-triazole
Solvent or Matrix Methanol
Molecular Weight (g/mol) 372.145
PubChem CID 80277
Quantity 1 mL
ChEBI CHEBI:83999
Chemical Name or Material Tetraconazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Tetraconazole 100ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.