missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethene in Methanol 10μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12910154 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12910154 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12910154 Supplier Fisher Chemical Supplier No. P853380
  • ONLINE EXCLUSIVE PRICE
    £40.60
    £36.69 / 10mL
    Save £3.91
    9% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 127-18-4
Physical Form Liquid
Molecular Formula C2Cl4
Concentration 10 μg/mL
MDL Number MFCD00000834
UN Number 1230
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES ClC(Cl)=C(Cl)Cl
IUPAC Name tetrachloroethene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 165.82
PubChem CID 31373
Quantity 10 mL
ChEBI CHEBI:17300
Chemical Name or Material Tetrachloroethene(Tetrachloroethylene)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Tetrachloroethylene 10ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.