missing translation for 'onlineSavingsMsg'
Learn More

Saliciylic acid in Isopropanol 2000μg/mL, Fisher Chemical™

Product Code. 12956893 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. Quantity unitSize
12956893 5 mL 5mL
1 options
This item is not returnable. View return policy

Product Code. 12956893

Brand: Fisher Chemical P878490

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 69-72-7
Analyte or Component Names Saliciylic acid in Isopropanol
Molecular Formula C7H6O3
Concentration 2000 μg/mL
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=C1O
Element Saliciylic Acid
IUPAC Name 2-hydroxybenzoic acid
Solvent or Matrix Isopropanol
Molecular Weight (g/mol) 138.12
PubChem CID 338
Quantity 5 mL
ChEBI CHEBI:16914
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.