missing translation for 'onlineSavingsMsg'
Learn More

Pyrene in DCM 100μg/mL, Fisher Chemical™

Product Code. 12906513 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
12906513 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 12906513

Brand: Fisher Chemical P850300

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 129-00-0
Physical Form Liquid
Molecular Formula C16H10
Concentration 100 μg/mL
InChI Key BBEAQIROQSPTKN-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
IUPAC Name pyrene
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 202.256
PubChem CID 31423
Quantity 1 mL
ChEBI CHEBI:39106
Chemical Name or Material Pyrene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.