missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Packaging | Am;poule |
| CAS | 24579-73-5 |
| Analyte or Component Names | Propamocarb free base in Methanol |
| Molecular Formula | C9H20N2O2 |
| Concentration | 100 μg/mL |
| InChI Key | WZZLDXDUQPOXNW-UHFFFAOYSA-N |
| SMILES | CCCOC(=O)NCCCN(C)C |
| Element | Propamocarb free base |
| IUPAC Name | propyl N-[3-(dimethylamino)propyl]carbamate |
| Solvent or Matrix | Methanol |
| Show More |
Safety and Handling
Product Identifier
- 1 ML Metabolite Propamocarb free base in Methanol
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: methanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
