missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Pentafluorophenol in DCM 100μg/mL, Fisher Chemical™

Product Code. 12996343 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12996343 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12996343 Supplier Fisher Chemical Supplier No. P847210

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 771-61-9
Molecular Formula C6HF5O
Concentration 100 μg/mL
MDL Number MFCD00002156
UN Number 1230
InChI Key XBNGYFFABRKICK-UHFFFAOYSA-N
SMILES OC1=C(F)C(F)=C(F)C(F)=C1F
IUPAC Name pentafluorophenol
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 184.07
PubChem CID 13041
Quantity 1 mL
Chemical Name or Material Pentafluorophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.