missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCB 30 in n-Hexane+C4221 400μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 35693-92-6 |
| Molecular Formula | C12H7Cl3 |
| Concentration | 400 μg/mL |
| UN Number | 1208 |
| InChI Key | MTLMVEWEYZFYTH-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | C1=CC=C(C=C1)C2=C(C=C(C=C2Cl)Cl)Cl |
| IUPAC Name | 1,3,5-trichloro-2-phenylbenzene |
| Solvent or Matrix | n-Hexane |
| Show More |
Safety and Handling
Product Identifier
- ORGANIC SINGLE COMPONENT STANDARD SOLUTION 400 ug/ml PCB 30 in n-Hexane (F131411)
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P321-Specific treatment (see on this label).
- P331-Do NOT induce vomiting.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction