missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mecoprop D3 in Acetone 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
10 mL
Packaging:
Ampule
Specifications
Packaging | Ampule |
Quantity | 10 mL |
CAS | 352431-15-3 |
Molecular Formula | C10H11ClO3 |
UN Number | 1090 |
Synonym | mecoprop-d3, 2-4-chloro-2,3,5-trideuterio-6-methylphenoxy propanoic acid, mechlorprop-d3, isocarnox-d3, compitox-d3, rankotex-d3, morogal-d3, okultin mp-d3, celatox cmpp-d3, anicon b-d3 |
InChI Key | WNTGYJSOUMFZEP-QGZYMEECSA-N |
SMILES | CC1=C(C=CC(=C1)Cl)OC(C)C(=O)O |
IUPAC Name | 2-(4-chloro-2,3,5-trideuterio-6-methylphenoxy)propanoic acid |
Molecular Weight (g/mol) | 217.663 |
Show More |
Safety and Handling
Product Identifier
- Standard solution Mecoprop D3 100ug/ml in acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'