missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Lindane in Methanol 10μg/mL, Fisher Chemical™

Product Code. 12986833 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12986833 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12986833 Supplier Fisher Chemical Supplier No. P827710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. Rückgaberichtlinie anzeigen

Spezifikation

Packaging Ampoule(s)
CAS 58-89-9
Molecular Formula C6H6Cl6
Concentration 10 μg/mL
MDL Number MFCD00135947
UN Number 1230
InChI Key JLYXXMFPNIAWKQ-UHFFFAOYSA-N
SMILES ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl
IUPAC Name 1,2,3,4,5,6-hexachlorocyclohexane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 290.81
PubChem CID 727
Quantity 10 mL
ChEBI CHEBI:24536
Chemical Name or Material Gamma-HCH (Lindane)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Lindane 10ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.