missing translation for 'onlineSavingsMsg'
Learn More

Hexachlorobenzene in Iso-octane 1000μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12966113 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12966113 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12966113 Supplier Fisher Chemical Supplier No. F117491
  • ONLINE EXCLUSIVE PRICE
    £80.25
    £62.22 / 1mL
    Save £18.03
    22% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 118-74-1
Molecular Formula C6Cl6
Concentration 1000 μg/mL
MDL Number MFCD00000540
UN Number 1262
InChI Key CKAPSXZOOQJIBF-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES ClC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl
IUPAC Name 1,2,3,4,5,6-hexachlorobenzene
Solvent or Matrix Isooctane
Molecular Weight (g/mol) 284.77
PubChem CID 8370
Quantity 1 mL
ChEBI CHEBI:5692
Chemical Name or Material Hexachlorobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • ORGANIC SINGLE COMPONENT STANDARD SOLUTION 1000 ug/ml Hexachlorobenzene in Iso-octane (F117491)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.