missing translation for 'onlineSavingsMsg'
Learn More

Glyphosate in water 10μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12926823 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12926823 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12926823 Supplier Fisher Chemical Supplier No. P827290
  • ONLINE EXCLUSIVE PRICE
    £48.90
    £48.00 / 10mL
    Save £0.90
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 1071-83-6
Molecular Formula C3H8NO5P
Concentration 10 μg/mL
MDL Number MFCD00055350
InChI Key XDDAORKBJWWYJS-UHFFFAOYSA-N
SMILES OC(=O)CNCP(O)(O)=O
IUPAC Name 2-[(phosphonomethyl)amino]acetic acid
Solvent or Matrix Water
Molecular Weight (g/mol) 169.07
PubChem CID 3496
Quantity 10 mL
ChEBI CHEBI:27744
Chemical Name or Material Glyphosate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.