missing translation for 'onlineSavingsMsg'
Learn More

Fluoranthene in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12966763 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12966763

Brand: Fisher Chemical P826200

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 206-44-0
Molecular Formula C16H10
Concentration 100 μg/mL
MDL Number MFCD00001184
InChI Key GVEPBJHOBDJJJI-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13
IUPAC Name fluoranthene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 202.26
PubChem CID 9154
Quantity 1 mL
ChEBI CHEBI:33083
Chemical Name or Material Fluoranthene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.