missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluazinam in Methanol 100μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule(s) |
| CAS | 79622-59-6 |
| Molecular Formula | C13H4Cl2F6N4O4 |
| Concentration | 100 μg/mL |
| MDL Number | MFCD00214168 |
| InChI Key | UZCGKGPEKUCDTF-UHFFFAOYSA-N |
| SMILES | [O-][N+](=O)C1=CC(=C(Cl)C(=C1NC1=NC=C(C=C1Cl)C(F)(F)F)[N+]([O-])=O)C(F)(F)F |
| IUPAC Name | 3-chloro-N-[3-chloro-2,6-dinitro-4-(trifluoromethyl)phenyl]-5-(trifluoromethyl)pyridin-2-amine |
| Solvent or Matrix | Methanol |
| Molecular Weight (g/mol) | 465.09 |
| Show More |
Safety and Handling
Product Identifier
- Standard solution Fluazinam 100ug/ml in methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- EUH208-Contains . May produce an allergic reaction.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P330-Rinse mouth.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction