missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate in Ethanol 10μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12946683 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12946683 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12946683 Supplier Fisher Chemical Supplier No. P824400
  • ONLINE EXCLUSIVE PRICE
    £38.85
    £38.15 / 10mL
    Save £0.70
    1% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 141-78-6
Molecular Formula C4H8O2
Concentration 10 μg/mL
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES CCOC(=O)C
IUPAC Name ethyl acetate
Solvent or Matrix Ethanol
Molecular Weight (g/mol) 88.106
PubChem CID 8857
Quantity 10 mL
ChEBI CHEBI:27750
Chemical Name or Material Ethyl Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.